missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calnexin Antibody (CANX/1543), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79835AF594
This item is not returnable.
View return policy
Description
Calnexin Monoclonal specifically detects Calnexin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Calnexin | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CANX | |
| Recombinant N-terminal fragment of human Calnexin protein (around aa 1-300) (exact sequence is proprietary) (Uniprot: P27824) | |
| 0.1 mL | |
| ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision | |
| This antibody recognizes a protein of 90kDa, which is identified as Calnexin. | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CANX/1543 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction