missing translation for 'onlineSavingsMsg'
Learn More

Calnexin Antibody (CANX/1541), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18408174 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408174 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408174 Supplier Novus Biologicals Supplier No. NBP254552AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Calnexin Monoclonal antibody specifically detects Calnexin in Human, Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calnexin
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CANX/1541
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
Host Species Mouse
Immunogen Recombinant N-terminal fragment of human Calnexin protein (around aa 1-300) (exact sequence is proprietary) (Uniprot: P27824)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline ER Markers, Neuroscience, Sensory Systems, Unfolded Protein Response, Vision
Primary or Secondary Primary
Gene ID (Entrez) 821
Target Species Human, Rat (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.