missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calnexin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB100-1974-0.025ml
This item is not returnable.
View return policy
Description
Calnexin Polyclonal specifically detects Calnexin in Human, Mouse, Rat, Hamster, Zebrafish samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Calnexin | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90 | |
| Rabbit | |
| 97 kDa | |
| 0.025 mL | |
| Neuroscience, Sensory Systems, Vision | |
| 821 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1:1000, Simple Western 1:50, Flow Cytometry, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:100 | |
| P35565 | |
| CANX | |
| A synthetic peptide made to a C-terminal portion of the rat Calnexin protein (between residues 550-591) [UniProt P35565] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Hamster | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction