missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calmodulin Antibody (2D1) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB120-2860JF669
This item is not returnable.
View return policy
Description
Calmodulin Monoclonal antibody specifically detects Calmodulin in Bacteria,Bovine,Chicken,Human,Mouse,Rat samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Calmodulin | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CALM, CALM2, CALM3, CALML2, calmodulin, calmodulin 1 (phosphorylase kinase, delta), CaM, CAM1, CAM2, CAM3, CAMB, CAMC, CAMI, CAMIII, DD132, PHKDCAM, phosphorylase kinase, delta subunit | |
| Calmodulin purified from Dictyostelium discoideum. | |
| 0.2 mL | |
| Cell Cycle and Replication, Neuroscience, Neurotransmission, Signal Transduction | |
| 801 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| 2D1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Bacteria, Bovine, Chicken, Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction