missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calmodulin Antibody (2D1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£169.00
Specifications
| Antigen | Calmodulin |
|---|---|
| Clone | 2D1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500, Flow Cytometry 2 μg/ 106 cells, ELISA 1:100 - 1:2000, Immunohistochemistry 1:20, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:20 |
| Applications | Western Blot |
Description
Calmodulin Monoclonal specifically detects Calmodulin in Human, Mouse, Rat, Bacteria, Bovine, Chicken samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Calmodulin | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication, Signal Transduction | |
| CALM, CALM2, CALM3, CALML2, calmodulin, calmodulin 1 (phosphorylase kinase, delta), CaM, CAM1, CAM2, CAM3, CAMB, CAMC, CAMI, CAMIII, DD132, PHKDCAM, phosphorylase kinase, delta subunit | |
| CALM1 | |
| IgG1 | |
| Protein A purified | |
| This detects calmodulin. It does not detect parvalbumin, tropinin, S-100, or myosin light chain kinase (MLCK). |
| 2D1 | |
| Western Blot 1:500, Flow Cytometry 2 μg/ 106 cells, ELISA 1:100 - 1:2000, Immunohistochemistry 1:20, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:20 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 801 | |
| Calmodulin purified from Dictyostelium discoideum. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title