missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calmodulin 1/2/3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102378
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Calmodulin1/2/3.
Calmodulin is a small, highly conserved calcium binding protein found in all eukaryotic cells. With the capacity to bind up to four calcium ions, this 17 kDa protein acts as an important intracellular receptor for regulatory calcium signals. As it binds calcium, calmodulin undergoes conformational changes which can increase its affinity for target proteins. It acts both directly, through interaction with key target enzymes, and indirectly, via specific kinases. Studies have found that calmodulin participates in the regulation of several biological processes including energy and biosynthetic metabolism, cell motility, exocytosis, cytoskeletal assembly, and intracellular modulation of both cAMP and calcium concentrations.
Specifications
| Calmodulin 1/2/3 | |
| Polyclonal | |
| Unconjugated | |
| CALM1 | |
| 1500001E21Rik; AI256814; AI327027; AI461935; AL024000; AL024017; Calm; CALM1; Calm2; CALM3; CALML2; calmodulin; calmodulin 1; Calmodulin 1 (phosphorylase kinase, delta); calmodulin 2; calmodulin 2 (phosphorylase kinase, delta); calmodulin 3; calmodulin 3 (phosphorylase kinase, delta); Calmodulin III; Calmodulin-1; Calmodulin-2; Calmodulin-3; Cam; CAM1; Cam2; CAM3; CaMA; CAMB; CAMC; CAMI; CAMII; CAMIII; CPVT4; DD132; epididymis secretory protein Li 72; HEL-S-72; LP7057 protein; LQT14; LQT15; PHKD; PHKD2; PHKD3; phosphorylase kinase delta; phosphorylase kinase subunit delta; phosphorylase kinase, delta subunit; prepro-calmodulin 1; prepro-calmodulin 2; prepro-calmodulin 3; R75142; RNCAMIII | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12313, 12314, 12315, 24242, 24244, 50663, 801, 805, 808 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P0DP23, P0DP24, P0DP25, P0DP26, P0DP27, P0DP28, P0DP29, P0DP30, P0DP31 | |
| CALM1, CALM2, CALM3 | |
| A synthesized peptide derived from human CALM1(Accession P0DP23), corresponding to amino acid residues V56-L106. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction