missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Calmodulin 2 Polyclonal antibody specifically detects Calmodulin 2 in Human samples. It is validated for ELISA
Specifications
Specifications
| Gene Alias | CALM, CALM1, CALM3, CALML2, calmodulin, calmodulin 2 (phosphorylase kinase, delta), CAM, CAM1, CAM2, CAM3, CAMB, CAMC, CAMII, CAMIII, FLJ99410, LP7057 protein, PHKD, PHKD2, phosphorylase kinase delta |
| Research Discipline | Cancer |
| Target Species | Human |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?