missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALML5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13059-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a novel calcium binding protein expressed in the epidermis and related to the calmodulin family of calcium binding proteins. Functional studies with recombit protein demonstrate it does bind calcium and undergoes a conformational change when it does so. Abundant expression is detected only in reconstructed epidermis and is restricted to differentiating keratinocytes. In addition, it can associate with transglutaminase 3, shown to be a key enzyme in the terminal differentiation of keratinocytes.Specifications
| CALML5 | |
| Polyclonal | |
| Unconjugated | |
| CALML5 | |
| CALML5, calmodulin like 5, Calmodulin like protein 5, Calmodulin like skin protein, CLSP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51806 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NZT1 | |
| CALML5 | |
| CALML5 Fusion Protein Ag3734 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction