missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CALML5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122174
This item is not returnable.
View return policy
Description
CALML5 Polyclonal Antibody for Western Blot, IHC (P), Flow
This gene encodes a novel calcium binding protein expressed in the epidermis and related to the calmodulin family of calcium binding proteins. Functional studies with recombinant protein demonstrate it does bind calcium and undergoes a conformational change when it does so. Abundant expression is detected only in reconstructed epidermis and is restricted to differentiating keratinocytes. In addition, it can associate with transglutaminase 3, shown to be a key enzyme in the terminal differentiation of keratinocytes.
Specifications
| CALML5 | |
| Polyclonal | |
| Unconjugated | |
| CALML5 | |
| call5; Calm4; CALML5; calmodulin 4; calmodulin like 5; calmodulin-like 5; calmodulin-like protein 5; Calmodulin-like skin protein; CLSP | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 51806 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9NZT1 | |
| CALML5 | |
| Recombinant protein Calmodulin Like Protein 5. The antigen corresponds to amino acid range 1-146 of the target protein. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction