missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALML5 Antibody (119), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89981B
This item is not returnable.
View return policy
Description
CALML5 Monoclonal specifically detects CALML5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CALML5 | |
| Monoclonal | |
| Biotin | |
| calmodulin-like 5, calmodulin-like protein 5, CLSPCalmodulin-like skin protein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CALML5 (AAH39172.1; Met1-Glu146). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 119 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 51806 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction