missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CALML3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568125
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
May function as a specific light chain of unconventional myosin-10 (MYO10), also enhances MYO10 translation, possibly by acting as a chaperone for the emerging MYO10 heavy chain protein. May compete with calmodulin by binding, with different affinities, to cellular substrates. [UniProt].
Specifications
| CALML3 | |
| Polyclonal | |
| Unconjugated | |
| CALML3 | |
| 2310068O22Rik; AI326174; CALML3; calmodulin like 3; calmodulin-like 3; calmodulin-like protein 3; Calmodulin-related protein NB-1; caM-like protein; CLP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 307100, 70405, 810 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P27482, Q5U206, Q9D6P8 | |
| CALML3 | |
| Synthesized peptide derived from human CALML3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction