Learn More
CALML3 (Human) Matched Antibody Pair, Abnova™
Matched Antibody Pair
Brand: Abnova H00000810-AP11.1Set
This item is not returnable.
View return policy
Specifications
| CALML3 | |
| Unconjugated | |
| CLP | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-CALML3 (100 ug) 2. Detection antibody: mouse monoclonal anti-CALML3, IgG2b Kappa (20 ug) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| calmodulin-like 3 | |
| CALML3 | |
| RUO | |
| 810 | |
| Human | |
| Ab Pair |
Product Content Correction
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu