missing translation for 'onlineSavingsMsg'
Learn More

Calmidazolium chloride, 99%, Thermo Scientific™

Product Code. 10295531 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10295531 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10295531 Supplier Thermo Scientific Acros Supplier No. 328140050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 57265-65-3
Melting Point 172°C to 176°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C31H23Cl7N2O
MDL Number MFCD00077679
Quantity 5 mg
Synonym calmidazolium chloride, 1-bis 4-chlorophenyl methyl-3-2-2,4-dichlorophenyl-2-2,4-dichlorobenzyloxy ethyl-1h-imidazolium chloride, 3-bis 4-chlorophenyl methyl-1-2-2,4-dichlorophenyl-2-2,4-dichlorophenyl methoxy ethyl imidazol-1-ium chloride, r 24571 chloride, methoxy ethyl-, chloride, calmidazolium chloride, solid, 1-bis p-chlorophenyl methyl-3-2,4-dichloro-beta-2,4-dichlorobenzyloxy phenethyl, 1-bis 4-chlorophenyl methyl-3-2,4-dichloro-, a-2,4-dichlorobenzyloxy phenethyl imidazolium chloride, 1-bis 4-chlorophenyl methyl-3-2,4-dichloro-beta-2,4-dichlorobenzyloxy phenethyl imidazolium chloride, 1-bis 4-chlorophenyl methyl-3-2-2,4-dichlorophenyl-2-2,4-dichlorophenyl methoxy ethyl imidazol-1-ium chloride
InChI Key YGEIMSMISRCBFF-UHFFFAOYSA-M
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)Cl)N3C=C[N+](=C3)CC(C4=C(C=C(C=C4)Cl)Cl)OCC5=C(C=C(C=C5)Cl)Cl)Cl.[Cl-]
IUPAC Name 1-[bis(4-chlorophenyl)methyl]-3-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazol-3-ium;chloride
Molecular Weight (g/mol) 687.688
PubChem CID 644274
Formula Weight 687.7
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material Calmidazolium chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.