missing translation for 'onlineSavingsMsg'
Learn More

California Petroleum Ether 2000 μg/mL in Triacetin, Dr. Ehrenstorfer

Product Code. 30325258
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325258 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325258 Supplier Dr. Ehrenstorfer Supplier No. DREGS09010406TN
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Petroleum ether
CAS 8032-32-4
Analyte or Component Names Petroleum ether
Shipping Condition Room Temperature
Concentration 2000 μg/mL
Certifications/Compliance ISO 17034
Synonym Ligroine, BP 2, BP 2 (solvent), Benzine (light petroleum distillate), Benzoline, Canadol, Ligroin, Naphtha, ligroine, Painters' naphtha, Petroleum ether, Refined solvent naphtha, Skellysolve F, Skellysolve G, V.M. and P. naphtha, V.M.&P. naphtha, Varnish makers' and painters' naphtha, Varnish makers' naphtha, Varnish makers' naphtha and painters' naphtha
Solvent or Matrix Triacetin
Quantity 5 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.