missing translation for 'onlineSavingsMsg'
Learn More

California Pesticide Class 2B Mixture 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30321542
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30321542 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 30321542

Brand: Dr. Ehrenstorfer DREGA09000669AL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexythiazox∣Imidacloprid∣Kresoxim-methyl∣Malathion∣Metalaxyl∣Methomyl∣Myclobutanil∣Naled∣Oxamyl∣Permethrin∣Phosmet∣Piperonyl butoxide∣Prallethrin∣Propiconazole∣Pyrethrins∣Pyridaben∣Quintozene∣Spinetoram J∣Spinosad∣Spiromesifen∣Spirotetramat∣Tebuconazole∣Thiamethoxam∣Trifloxystrobin
CAS 78587-05-0, 138261-41-3, 143390-89-0, 121-75-5, 57837-19-1, 16752-77-5
Analyte or Component Names Hexythiazox∣Imidacloprid∣Kresoxim-methyl∣Malathion∣Metalaxyl∣Methomyl∣Myclobutanil∣Naled∣Oxamyl∣Permethrin∣Phosmet∣Piperonyl butoxide∣Prallethrin∣Propiconazole∣Pyrethrins∣Pyridaben∣Quintozene∣Spinetoram J∣Spinosad∣Spiromesifen∣Spirotetramat∣Tebuconazole∣Thiamethoxam∣Trifloxystrobin
Shipping Condition Ice pack (-20°C)
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Hexythiazox 100 μg/mL∣Imidacloprid 100 μg/mL∣Kresoxim-methyl 100 μg/mL∣Malathion 100 μg/mL∣Metalaxyl 100 μg/mL∣Methomyl 100 μg/mL∣Myclobutanil 100 μg/mL∣Naled 100 μg/mL∣Oxamyl 100 μg/mL∣Permethrin 100 μg/mL∣Phosmet 100 μg/mL∣Piperonyl butoxide 100 μg/mL∣Prallethrin 100 μg/mL∣Propiconazole 100 μg/mL∣Pyrethrins 100 μg/mL∣Pyridaben 100 μg/mL∣Quintozene 100 μg/mL∣Spinetoram J 100 μg/mL∣Spinosad 100 μg/mL∣Spiromesifen 100 μg/mL∣Spirotetramat 100 μg/mL∣Tebuconazole 100 μg/mL∣Thiamethoxam 100 μg/mL∣Trifloxystrobin 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.