missing translation for 'onlineSavingsMsg'
Learn More

Calicheamicin, MedChemExpress

Product Code. p-200096976
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16491829

Brand: Medchem Express HY19609

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Calicheamicin, an antitumor antibiotic, is a cytotoxic agent that causes double-strand DNA breaks. Calicheamicin is a DNA synthesis inhibitor.

Calicheamicin, an antitumor antibiotic, is a cytotoxic agent that causes double-strand DNA breaks. Calicheamicin is a DNA synthesis inhibitor.

Specifications

Chemical Name or Material Calicheamicin
Physical Form Solid
CAS 108212-75-5
Color Light Yellow
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C55H74IN3O21S4
For Use With (Application) Cancer-programmed cell death
Synonym Calicheamicin γ1
Solubility Information DMSO : 25 mg/mL (18.27 mM; Need ultrasonic)
SMILES O[C@](CC1=O)(C#C/C=C\C#C2)/C(C([C@H]2O[C@@](O[C@H](C)[C@@H](NO[C@H](O[C@H](C)[C@H]3SC(C(C(C)=C(I)C(O[C@H](O[C@@H](C)[C@H](O)[C@H]4OC)[C@@H]4O)=C5OC)=C5OC)=O)C[C@@H]3O)[C@@H]6O)([H])[C@@H]6O[C@@](OC[C@@H]7NCC)([H])C[C@@H]7OC)=C1NC(OC)=O)=C\CSSSC
Molecular Weight (g/mol) 1368.35
Formula Weight 1368.35
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.