missing translation for 'onlineSavingsMsg'
Learn More

Calicheamicin, MedChemExpress

Product Code. 16491829
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491829 1 mg 1mg
16480278 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491829 Supplier Medchem Express Supplier No. HY19609

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Calicheamicin, an antitumor antibiotic, is a cytotoxic agent that causes double-strand DNA breaks. Calicheamicin is a DNA synthesis inhibitor.

Calicheamicin, an antitumor antibiotic, is a cytotoxic agent that causes double-strand DNA breaks. Calicheamicin is a DNA synthesis inhibitor.

Specifications

Chemical Name or Material Calicheamicin
Physical Form Solid
CAS 108212-75-5
Color Light Yellow
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C55H74IN3O21S4
For Use With (Application) Cancer-programmed cell death
Synonym Calicheamicin γ1
Solubility Information DMSO : 25 mg/mL (18.27 mM; Need ultrasonic)
SMILES O[C@](CC1=O)(C#C/C=C\C#C2)/C(C([C@H]2O[C@@](O[C@H](C)[C@@H](NO[C@H](O[C@H](C)[C@H]3SC(C(C(C)=C(I)C(O[C@H](O[C@@H](C)[C@H](O)[C@H]4OC)[C@@H]4O)=C5OC)=C5OC)=O)C[C@@H]3O)[C@@H]6O)([H])[C@@H]6O[C@@](OC[C@@H]7NCC)([H])C[C@@H]7OC)=C1NC(OC)=O)=C\CSSSC
Molecular Weight (g/mol) 1368.35
Formula Weight 1368.35
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.