missing translation for 'onlineSavingsMsg'
Learn More

Xylem Calibration Oil 5 x 5 mL Bottles (UKAS/NIST)

Product Code. 30484450
Change view
Click to view available options
Refractive Index:
1.46666
1.55768
Unit Size:
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Refractive Index Unit Size
30484450 1.46666 25mL
30484456 1.55768 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30484450 Supplier Xylem Supplier No. 90525
  • ONLINE EXCLUSIVE PRICE
    £181.00
    £145.96 / 25mL
    Save £35.04
    19% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Calibration oil for calibration of refractometers.

  • Calibration Oil
  • For Refractometers
TRUSTED_SUSTAINABILITY

Specifications

Type Calibration Fluid
Refractive Index 1.46666
For Use With (Equipment) Refractometer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.