missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Calibration Kit for CyberScan TB1000 Turbidity Meter

Product Code. 10424784
Click to view available options
For Use With (Equipment):
CyberScan TB 1000
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10424784

Brand: Thermo Scientific™ ECTBDWLCALKT

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The CyberScan TB1000 series turbidity meters and calibration kits are designed to meet the stringent requirements of today's scientists.

The CyberScan TB1000 series turbidity meters and calibration kits are designed to meet the stringent requirements of today's scientists. TB1000 series meet the performance criteria specified by USEPA method 180.1 for NTU measurement and ISO 7027 (IR version). Ideal for beverage production, drinking water, waste water treatment, petrochemical and electroplating applications.

Features:

  • Calibration Points - 0.02, 10.0, 1000 NTU
  • Used with TB1000 Meter ECTBDW1000
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) CyberScan TB 1000
Description Calibration Kit Set for CyberScan TB1000 ECTBDW1000, Calibration points - 0.02, 10.0, 1000 NTU
compliance-icons
Product Identifier
  • Turbidity standards
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : styrene divinyl benzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.