Learn More
BINDER™ Calibration Certificate, Tempurature and Humidity
Calibration certificate, temperature and humidity - Measurement in center of chamber at 25°C / 60% RH or at specified test values
Brand:  BINDER™ 8012-1187
Additional Details : Weight : 0.00200kg
 View more versions of this product
Product Code. 15922962
  • £446.00 / Each
Estimated Shipment: 21-08-2024
to see stock.
Description
Description
  • One humidity point of measurement
  • Including certificate
  • Humidity measurement point in center of chamber
  • Temperature (according to DIN12880) and humidity measurement at 25°C / 60% RH or at specified values
Specifications
Specifications
BINDER Chambers (Models MKFT 115, MKFT 240, MKFT 720)
Calibration Certificate (Temperature and Humidity)
Temperature and Humidity Measurement
Calibration certificate, temperature and humidity - Measurement in center of chamber at 25°C / 60% RH or at specified test values
1 measuring point