missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALHM3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | CALHM3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
CALHM3 Polyclonal specifically detects CALHM3 in Human samples. It is validated for Western Blot.Specifications
| CALHM3 | |
| Polyclonal | |
| Rabbit | |
| bA225H22.7, calcium homeostasis modulator 3, calcium homeostasis modulator protein 3, FAM26A, family with sequence similarity 26, member A | |
| CALHM3 | |
| IgG | |
| 38 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 119395 | |
| Synthetic peptides corresponding to FAM26A The peptide sequence was selected from the C terminal of FAM26A. Peptide sequence RSELQARGLRRGNAGRRLELPAVPEPPEGLDSGSGKAHLRAISSREQVDR. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title