missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALHM1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | CALHM1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18657642
|
Novus Biologicals
NBP2-92818-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18676572
|
Novus Biologicals
NBP2-92818-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CALHM1 Polyclonal antibody specifically detects CALHM1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CALHM1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 255022 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| calcium homeostasis modulator 1, FAM26Ccalcium homeostasis modulator protein 1, family with sequence similarity 26, member C, MGC39514, MGC39617, Protein FAM26C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 202-346 of human CALHM1 (NP_001001412.3). VRSVRPCFTQAAFLKSKYWSHYIDIERKLFDETCTEHAKAFAKVCIQQFFEAMNHDLELGHTHGTLATAPASAAAPTTPDGAEEEREKLRGITDQGTMNRLLTSWHKCKPPLRLGQEEPPLMGNGWAGGGPRPPRKEVATYFSKV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title