missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calhex 231 hydrochloride, Tocris Bioscience™
Negative allosteric modulator of calcium-sensing receptor (CaSR)
£241.25 - £905.25
Chemical Identifiers
CAS | 652973-93-8 |
---|---|
Molecular Formula | C25H27ClN2O |
Molecular Weight (g/mol) | 406.954 |
InChI Key | YTFUQWWKTIWYEY-VEYWIMSWSA-N |
Synonym | benzamide,4-chloro-n-1s,2s-2-1r-1-1-naphthalenyl ethyl amino cyclohexyl |
PubChem CID | 97103734 |
IUPAC Name | 4-chloro-N-[(1S,2R)-2-[[(1R)-1-naphthalen-1-ylethyl]amino]cyclohexyl]benzamide |
SMILES | CC(C1=CC=CC2=CC=CC=C21)NC3CCCCC3NC(=O)C4=CC=C(C=C4)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18045252
|
Tocris Bioscience
4387/10 |
10 mg |
£241.25
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18035122
|
Tocris Bioscience
4387/50 |
50 mg |
£905.25
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Negative allosteric modulator of the calcium sensing receptor (CaSR). Blocks increases in [3H]inositol phosphate levels elicited by wild-type hCasR activation (IC50= 0.39 μM in transiently transfected HEK293 cells).Chemical Identifiers
652973-93-8 | |
406.954 | |
benzamide,4-chloro-n-1s,2s-2-1r-1-1-naphthalenyl ethyl amino cyclohexyl | |
4-chloro-N-[(1S,2R)-2-[[(1R)-1-naphthalen-1-ylethyl]amino]cyclohexyl]benzamide |
C25H27ClN2O | |
YTFUQWWKTIWYEY-VEYWIMSWSA-N | |
97103734 | |
CC(C1=CC=CC2=CC=CC=C21)NC3CCCCC3NC(=O)C4=CC=C(C=C4)Cl |
Specifications
652973-93-8 | |
benzamide,4-chloro-n-1s,2s-2-1r-1-1-naphthalenyl ethyl amino cyclohexyl | |
CC(C1=CC=CC2=CC=CC=C21)NC3CCCCC3NC(=O)C4=CC=C(C=C4)Cl | |
406.954 | |
443.41 | |
Calhex 231 hydrochloride |
C25H27ClN2O | |
YTFUQWWKTIWYEY-VEYWIMSWSA-N | |
4-chloro-N-[(1S,2R)-2-[[(1R)-1-naphthalen-1-ylethyl]amino]cyclohexyl]benzamide | |
97103734 | |
>98% |
Safety and Handling
Recommended Storage : Store at -20°C