missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calgranulin B Mouse, Clone: CAGB/426, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 6280-MSM2-P1
This item is not returnable.
View return policy
Specifications
| Calgranulin B | |
| Cytoplasmic | |
| CAGB/426 | |
| Unconjugated | |
| NP_002956 | |
| Mouse | |
| 14kDa | |
| Protein A/G Purified | |
| RUO | |
| 6280 | |
| IgM κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 1mM PBS with 0.05% BSA and 0.05% azide. | |
| S100A9 | |
| Recombinant human CAGB protein | |
| Tonsil, lymph node, or spleen | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction