missing translation for 'onlineSavingsMsg'
Learn More

Calflo E, Honeywell Fluka™

Product Code. p-7145729 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
4 x 250 g
Unit Size:
250g
Pack of 4
This item is not returnable. View return policy

Product Code. 16635833

Brand: Honeywell Fluka 34316H250G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

PESTANAL™, for pesticide residue analysis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-95-2
PubChem CID 14941

Specifications

Physical Form Solid
Quantity 250 g
Packaging Plastic Bottle
Suitability Max. 50 ppb
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.