missing translation for 'onlineSavingsMsg'
Learn More

Calflo E, Honeywell Fluka™

Product Code. 16645833 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
4 x 250 g
Unit Size:
250g
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16645833 4 x 250 g Pack of 4
16635833 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16645833 Supplier Honeywell Fluka Supplier No. 34316H4X250G
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PESTANAL™, for pesticide residue analysis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-95-2
PubChem CID 14941

Specifications

Physical Form Solid
Quantity 4 x 250 g
Packaging Plastic Bottle
Suitability Max. 50 ppb
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.