missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Caldesmon Recombinant Rabbit Monoclonal Antibody (24GB4735)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557349
This item is not returnable.
View return policy
Description
Caldesmon Recombinant Monoclonal Antibody for Western Blot, Flow
Caldesmon is a developmentally regulated protein involved in smooth muscle and non muscle contraction. Two closely related variants of human caldesmon have been identified which differ in their electrophorectic mobility and cellular distribution. The h caldesmon variant (120 - 150 kDa) is predominantly expressed in smooth muscle whereas l caldesmon (70 - 80 kDa) is found in non-muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle.
Specifications
| Caldesmon | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Cald1 | |
| 4833423D12Rik; AI195384; AV071549; AW536160; C920027I18Rik; CAD; Cald1; Caldesmon; caldesmon 1; CDM; HCAD; H-CAD; h-caldesmon; LCAD; L-CAD; L-caldesmon; MGC21352; NAG22; Non-muscle caldesmon; OTTHUMP00000207950; OTTHUMP00000207953; OTTHUMP00000207954; OTTHUMP00000207955; OTTHUMP00000214473; smooth muscle caldesmon; testis secretory sperm-binding protein Li 227n | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 109624, 25687, 800 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4735 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q05682, Q62736 | |
| Cald1 | |
| A synthesized peptide derived from human Caldesmon (aa 782-793). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction