missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Caldesmon Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582584
This item is not returnable.
View return policy
Description
Immunogen sequence: MTHKLKHTEN TFSRPGGRAS VDTKEAEGAP QVEAGKRLEE LRRRRGETES EEFEKLKQKQ QEAALELEEL KKKREERRKV LEEEEQRRKQ EEADRKLREE EEKRRLKEEI ERRRAEAAEK RQKMPEDGLS DD Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 93%.
Caldesmon is a developmentally regulated protein involved in smooth muscle and non muscle contraction. Two closely related variants of human caldesmon have been identified which differ in their electrophorectic mobility and cellular distribution. The h caldesmon variant (120 - 150 kDa) is predominantly expressed in smooth muscle whereas l caldesmon (70 - 80 kDa) is found in non-muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle.
Specifications
| Caldesmon | |
| Polyclonal | |
| Unconjugated | |
| Cald1 | |
| 4833423D12Rik; AI195384; AV071549; AW536160; C920027I18Rik; CAD; Cald1; Caldesmon; caldesmon 1; CDM; HCAD; H-CAD; h-caldesmon; LCAD; L-CAD; L-caldesmon; MGC21352; NAG22; Non-muscle caldesmon; OTTHUMP00000207950; OTTHUMP00000207953; OTTHUMP00000207954; OTTHUMP00000207955; OTTHUMP00000214473; smooth muscle caldesmon; testis secretory sperm-binding protein Li 227n | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 800 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q05682 | |
| Cald1 | |
| Recombinant protein corresponding to Human CALD1. Recombinant protein control fragment (Product #RP-89610). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction