missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Caldesmon HMW Monoclonal Antibody (h-CALD)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA511626
This item is not returnable.
View return policy
Description
MA5-11626 targets Caldesmon HMW in IHC (P), IF and WB applications and shows reactivity with human and mouse samples. The MA5-11626 immunogen is crude human uterus extract.
Caldesmon is a developmentally regulated protein involved in smooth muscle and non muscle contraction. Two closely related variants of human caldesmon have been identified which differ in their electrophorectic mobility and cellular distribution. The h caldesmon variant (120 - 150 kDa) is predominantly expressed in smooth muscle whereas l caldesmon (70 - 80 kDa) is found in non-muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle.
Specifications
| Caldesmon HMW | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide; pH 7.4 | |
| Q05682, Q62736 | |
| Cald1 | |
| Crude human uterus extract. | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| h-CALD | |
| Unconjugated | |
| Cald1 | |
| 4833423D12Rik; AI195384; AV071549; AW536160; C920027I18Rik; CAD; Cald1; Caldesmon; caldesmon 1; CDM; HCAD; H-CAD; h-caldesmon; LCAD; L-CAD; L-caldesmon; MGC21352; NAG22; Non-muscle caldesmon; OTTHUMP00000207950; OTTHUMP00000207953; OTTHUMP00000207954; OTTHUMP00000207955; OTTHUMP00000214473; smooth muscle caldesmon; testis secretory sperm-binding protein Li 227n | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 109624, 25687, 800 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction