missing translation for 'onlineSavingsMsg'
Learn More

Caldesmon/CALD1 Mouse anti-Human, Clone: rCALD1/820, Novus Biologicals™

Product Code. 18372463 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372463 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18372463 Supplier Novus Biologicals Supplier No. NBP30736720UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Caldesmon/CALD1 Monoclonal specifically detects Caldesmon/CALD1 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caldesmon/CALD1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rCALD1/820
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 0.25-0.5 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias CAD, caldesmon, caldesmon 1, CDMH-CAD, HCAD, LCAD, L-CAD, MGC21352, NAG22
Host Species Mouse
Immunogen Recombinant full-length human Caldesmon/CALD1 protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 800
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.