missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caldesmon/CALD1 Antibody (h-CALD), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47816MFV450
This item is not returnable.
View return policy
Description
Caldesmon/CALD1 Monoclonal antibody specifically detects Caldesmon/CALD1 in Human samples. It is validated for Flow Cytometry
Specifications
| Caldesmon/CALD1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| h-CALD | |
| Flow Cytometry | |
| CAD, caldesmon, caldesmon 1, CDMH-CAD, HCAD, LCAD, L-CAD, MGC21352, NAG22 | |
| Crude human uterus extract | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Phospho Specific | |
| 800 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction