missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAPS2 Monoclonal Antibody (3C6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524499
This item is not returnable.
View return policy
Description
Peptide Sequence: YRKSYVRKAF MKLDFNKSGS VPIINIRKCY CAKKHSQVIS GHSTEEEIKS SFLETLKVAC SKSDEVSYGE FEDYYEGLSI GIVDDEDFVN ILRTPWGI.
Calcyphosine-2 is a calcium-binding protein with 2 EF-hand motifs (Wang et al., 2002 [PubMed 11846421]).
Specifications
| CAPS2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9BXY5 | |
| CAPS2 | |
| CAPS2 (NP_115995.1, 285 a.a. ~ 382 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Western Blot | |
| 3C6 | |
| Unconjugated | |
| CAPS2 | |
| calcyphosin 2; calcyphosin-2; calcyphosine 2; calcyphosine-2; calcyphosphine 2; CAPS2; D630005B03Rik; UG0636c06 | |
| Mouse | |
| Protein A | |
| RUO | |
| 84698 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction