missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Calconcarboxylic acid, indicator grade
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Melting Point | 300°C |
| Color | Black to Purple |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Quantity | 50 g |
| Solubility Information | Solubility in water: slightly soluble. Other solubilities: soluble in methanol and ethanol |
| Formula Weight | 438.4 |
| Grade | Extra Pure |
| Physical Form | Powder |
| Chemical Name or Material | Calconcarboxylic acid |
Safety and Handling
Product Identifier
- Calconcarboxylic acid, indicator grade
- Warning
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?