missing translation for 'onlineSavingsMsg'
Learn More

Calconcarboxylic Acid, Solstice

Product Code. 15691940 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15691940 1 g Glass Bottle 1g
15681940 10 g Plastic Bottle 10g
15601950 50 g Plastic Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15691940 Supplier Honeywell-Fluka Supplier No. 212601G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

for complexometry

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 3737-95-9
Molecular Formula C21H14N2O7S
Molecular Weight (g/mol) 438.41
MDL Number MFCD00004078
InChI Key ULIVOAKVRBXKKS-PYCFMQQDSA-N
PubChem CID 5895210
IUPAC Name 3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C=C(C(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)O)O)C(=O)O

Tekniske data

Melting Point 300°C
Packaging Glass Bottle
UN Number NONH for all modes of transport
Beilstein 727492
Color Violet
Quantity 1 g
Physical Form Powder
Formula Weight 438.41g/mol
Chemical Name or Material Calconcarboxylic Acid, For complexometry
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.