missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Calcon™, pure

Product Code. p-178142 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10722631 25 g Glass bottle 25g
10722821 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10722631 Supplier Thermo Scientific Chemicals Supplier No. 202440250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2538-85-4 | C20H13N2NaO5S | 416.383 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2538-85-4
Molecular Formula C20H13N2NaO5S
Molecular Weight (g/mol) 416.383
MDL Number MFCD00004069
InChI Key YJUGDMOGPJJWDG-UHFFFAOYSA-M
Synonym C.I. 15705, Eriochrome™ Blue Black R, Mordant Black 17, Zinchrome R
PubChem CID 23702480
IUPAC Name sodium;4-[(2-hydroxynaphthalen-1-yl)hydrazinylidene]-3-oxonaphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C=CC(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)[O-])O.[Na+]

Specifications

Beilstein 16, 297
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in most organic solvents
Infrared Spectrum Authentic
Formula Weight 416.39
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Black to Brown
Quantity 25 g
Physical Form Powder
Chemical Name or Material Calcon™
Show More Show Less
Product Identifier
  • Calcon]TS

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.