missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CALCOCO1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558337
This item is not returnable.
View return policy
Description
Immunogen sequence: EAEDEKSVLM AAVQSGGEEA NLLLPELGSA FYDMASGFTV GTLSETSTGG PATPTWKECP ICKERFPAES DKDALEDHMD GHFF Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.
CALCOCO1 (calcium-binding and coiled-coil domain-containing protein 1), also known as cocoa, calphoglin, sarcoma antigen NY-SAR-3 or coiled-coil coactivator protein, is a 691 amino acid protein that shuttles between the cytoplasm and nucleus and functions as coactivator for aryl hydrocarbon and nuclear receptors. A member of the CALCOCO family, CALCOCO1 is forms a calphoglin complex with PPA1 and PGM 1 and contains multiple functional domains through which it acts as a component of both the androgen signaling pathway and the Wnt/beta-catenin signaling pathway.
Specifications
| CALCOCO1 | |
| Polyclonal | |
| Unconjugated | |
| Calcoco1 | |
| 1810009B06Rik; calcium binding and coiled coil domain 1; calcium binding and coiled-coil domain 1; calcium-binding and coiled-coil domain-containing protein 1; Calcoco1; Calphoglin; Cocoa; Coiled-coil coactivator protein; coiled-coil leucine zipper coactivator 1; coiled-coil transcriptional coactivator; Gcap11; granule cell antiserum positive 11; GRIP1-interacting protein; inorganic pyrophosphatase activator; KIAA1536; MGC128949 protein; mKIAA1536; PP13275; Sarcoma antigen NY-SAR-3; UNQ2436/PRO4996 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57658 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P1Z2 | |
| Calcoco1 | |
| Recombinant protein corresponding to Human CALCOCO1. Recombinant protein control fragment (Product #RP-97620). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction