missing translation for 'onlineSavingsMsg'
Learn More

Calcium zirconium oxide, 99.2% (metals basis), Thermo Scientific Chemicals

Product Code. 11391709
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391709 250 g 250g
11301719 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11391709 Supplier Thermo Scientific Alfa Aesar Supplier No. 012344.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Calcium zirconium oxide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12013-47-7
Molecular Formula CaO3Zr
Molecular Weight (g/mol) 179.299
MDL Number MFCD00015982
InChI Key BJJVDFHQHSBAFC-UHFFFAOYSA-N
Synonym calcium zirconate, calcium oxozirconiumbis olate, calcium;dioxido oxo zirconium, calcium 2+ oxozirconiumbis olate
PubChem CID 16212531
IUPAC Name calcium;dioxido(oxo)zirconium
SMILES [O-][Zr](=O)[O-].[Ca+2]

Specifications

Melting Point 2550°C
Physical Form Powder
Quantity 250 g
Solubility Information Soluble in Nitric acid.
Formula Weight 179.3
Percent Purity 99.2%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Calcium zirconium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.