missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium turnings, 98% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.54 |
Assay Percent Range | 98% (metals basis) |
Quantity | 10 g |
Formula Weight | 40.078 |
Form | Turnings |
Chemical Name or Material | Calcium |
Safety and Handling
Product Identifier
- Calcium turnings
- Danger
- Substance which\, in contact with water\, emits flammable gases Category 2
- H261-In contact with water releases flammable gases.
- P332-If skin irritation occurs:.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P233-Keep container tightly closed.
- P370+P378-In case of fire: Use to extinguish.
- P402+P404-Store in a dry place. Store in a closed container.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'