missing translation for 'onlineSavingsMsg'
Learn More

Calcium tungsten oxide, 99.78% (metals basis)

Product Code. 11353827
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353827 250 g 250g
11363827 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353827 Supplier Thermo Scientific Alfa Aesar Supplier No. 035669.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-75-2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-75-2
MDL Number MFCD00010913

Specifications

Melting Point 1620°C
Physical Form Powder
Quantity 250 g
Merck Index 14,1712
Formula Weight 287.93
Percent Purity 99.78%
Assay (metals basis)
Chemical Name or Material Calcium tungsten oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.