missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium tungsten oxide, 98%, Thermo Scientific Chemicals
£35.80 - £261.00
Chemical Identifiers
CAS | 7790-75-2 |
---|---|
MDL Number | MFCD00010913 |
Synonym | Calcium tungstate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11312138
|
Thermo Scientific Alfa Aesar
013051.14 |
25 g |
£35.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11302138
|
Thermo Scientific Alfa Aesar
013051.22 |
100 g |
£70.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11392128
|
Thermo Scientific Alfa Aesar
013051.36 |
500 g |
£261.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium tungsten oxide is used as a luminophore.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7790-75-2 | |
MFCD00010913 | |
Calcium tungstate |
Specifications
1620°C | |
Powder | |
14,1712 | |
Insoluble in H2O; Decomposes by hot HCl,HNO3 | |
98% |
7790-75-2 | |
MFCD00010913 | |
Calcium tungstate | |
287.93 | |
Calcium tungsten oxide |
Safety and Handling
missing translation for 'einecsNumber' : 232-219-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only