missing translation for 'onlineSavingsMsg'
Learn More

Calcium thiosulfate, pure, 30-50% solution in water

Product Code. p-4813390 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
This item is not returnable. View return policy

Product Code. 12655117

Brand: Thermo Scientific Acros 447870010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 10124-41-1 | CaO3S2 | 152.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium thiosulfate
CAS 10124-41-1, 7732-18-5
Quantity 1 L
Molecular Formula CaO3S2
Linear Formula CaS2O3
Synonym calcium thiosulfate, tecesal, calcium thiosulphate, calcium hyposulfite, caswell no. 153, unii-2io10o728e, calcium thiosulfate ca s2o3, epa pesticide chemical code 080101, thiosulfuric acid h2s2o3 , calcium salt 1:1, thiosulfuric acid, calcium salt 1:1
Solubility Information Solubility in water: soluble
InChI Key FAYYUXPSKDFLEC-UHFFFAOYSA-L
SMILES [O-]S(=O)(=S)[O-].[Ca+2]
IUPAC Name calcium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
Molecular Weight (g/mol) 152.21
PubChem CID 24964
Formula Weight 152.21
Grade Pure
Packaging Glass bottle
Specific Gravity 1.245
Density 1.2450g/mL
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.