missing translation for 'onlineSavingsMsg'
Learn More

Calcium Thioglycolate Trihydrate, ≥98.0% (RT), Honeywell™ Fluka™

Product Code. 15671940 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15671940 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15671940 Supplier Honeywell-Fluka Supplier No. 21250500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Crystallized,≥98.0% (RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 65208-41-5
Color Beige
Physical Form Crystallized Powder
Quantity 500 g
Molecular Formula C2H10CaO5S
Linear Formula -SCH2COOCa+ · 3H2O
MDL Number MFCD00150707
UN Number NONH for all modes of transport
Beilstein 3755791
Synonym calcium,2-sulfidoacetate,trihydrate, thioglycolic acid calcium trihydrate
InChI Key GDECCIJZCWLFSB-UHFFFAOYSA-N
SMILES O.O.O.[Ca].OC(=O)CS
IUPAC Name 2-sulfanylacetic acid calcium trihydrate
Molecular Weight (g/mol) 186.24
PubChem CID 131882808
Formula Weight 185.23g/mol
Percent Purity ≥98.0% (RT)
Packaging Glass bottle
Chemical Name or Material Calcium Thioglycolate Trihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.