missing translation for 'onlineSavingsMsg'
Learn More

Calcium sulfide, 99.9% (metals basis excluding Sr), Sr typically 500 ppm

Product Code. 11322218
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322218 5 g 5g
11302218 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322218 Supplier Thermo Scientific Alfa Aesar Supplier No. 013121.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20548-54-3 | CaS | 72.14 g/mol

It is used as luminous paint, depilatory, lubricant additive, ore dressing and flotation agent and as an arsenic-free hydrogen sulfide. It is also used as a depilatory in the tanning industry and in cosmetics, in homeopathy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20548-54-3
Molecular Formula CaS
Molecular Weight (g/mol) 72.14
MDL Number MFCD00015990
InChI Key AGVJBLHVMNHENQ-UHFFFAOYSA-N
PubChem CID 30182
IUPAC Name calcium sulfanediide
SMILES [S--].[Ca++]

Specifications

Melting Point (decomposition)
Physical Form Powder
Quantity 5 g
UN Number UN3134
Merck Index 14,1707
Solubility Information Slightly soluble in water. Insoluble in alcohol
Formula Weight 72.14
Percent Purity 99.9%
Odor Stench
Sensitivity Moisture sensitive
Density 2.5 g/mL
Assay (metals basis excluding Sr), Sr typically 500ppm
Chemical Name or Material Calcium sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.