missing translation for 'onlineSavingsMsg'
Learn More

Calcium Sulfate, Drierite™, Honeywell™ Fluka™

Product Code. 15626950 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
454 g
Packaging:
Glass Bottle
Unit Size:
454g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15626950 454 g Glass Bottle 454g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15626950 Supplier Honeywell-Fluka Supplier No. 737828454G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

With indicator, 10-20 mesh, Drierite™ is a trademark of W. A. Hammond Drierite Company, Ltd.

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Melting Point 1450°C
Physical Form Solid
CAS 7778-18-9
Material Calcium Sulfate
Quantity 454 g
Molecular Formula CaO4S
Linear Formula CaSO4
MDL Number MFCD00010912
UN Number UN3077
Packaging Glass Bottle
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
SMILES [Ca++].[O-]S([O-])(=O)=O
IUPAC Name calcium sulfate
Molecular Weight (g/mol) 136.13
PubChem CID 24497
ChEBI CHEBI:31346
Formula Weight 136.14g/mol
Chemical Name or Material Calcium sulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.