missing translation for 'onlineSavingsMsg'
Learn More

Regucalcin Antibody - BSA Free, R&D Systems™

Product Code. 18609541 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18609541 0.02 mL 0.02mL
18692052 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18609541 Supplier Novus Biologicals Supplier No. NBP2948510.02ml
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

Regucalcin Polyclonal specifically detects Regucalcin in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Regucalcin
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias EC 3.1.1.17, Gluconolactonase, GNL, RCsenescence marker protein-30, regucalcin (senescence marker protein-30), Senescence marker protein 30, SMP-30, SMP30regucalcin
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-299 of human RGN (NP_004674.1). MSSIKIECVLPENCRCGESPVWEEVSNSLLFVDIPAKKVCRWDSFTKQVQRVTMDAPVSSVALRQSGGYVATIGTKFCALNWKEQSAVVLATVDNDKKNNRFNDGKVDPAGRYFAGTMAEETAPAVLERHQGALYSLFPDHHVKKYFDQVDISNGLDWSLDHKIFYYIDSLSYSVDAFDYDLQTGQISNRRSVYKLEKEEQIPDGMCIDAEGKLWVACYNGGRVIRLDPVTGKRLQTVKLPVDKTTSCCFGGKNYSEMYVTCARDGMDPEGLLRQPEAGGIFKITGLGVKGIAPYSYAG
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9104
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.