missing translation for 'onlineSavingsMsg'
Learn More

Calcium sulfate dihydrate, Puriss., meets analytical spec. of NF, E 516, 99.0-101.0% (based on anhydrous substance), Honeywell Fluka™
BIOPHARMA

Product Code. p-7111747 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
25 kg
6 x 1 kg
Unit Size:
1kg
25kg
Pack of 6
This item is not returnable. View return policy

Product Code. 15631100

Brand: Honeywell-Fluka 120561KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number MFCD00149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
Synonym calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
PubChem CID 24928
ChEBI CHEBI:32583
IUPAC Name calcium;sulfate;dihydrate
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

Quantity 1 kg
Linear Formula CaSO4 · 2H2O
UN Number NONH for all modes of transport
Formula Weight 172.17g/mol
Percent Purity 99 to 101% (based on anhydrous substance)
Grade puriss.
Chemical Name or Material Calcium Sulfate Dihydrate, Meets analytical specification of NF
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.