missing translation for 'onlineSavingsMsg'
Learn More

Calcium sulfate, 99%, pure, anhydrous

Product Code. 11924251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
10kg
2.5kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11924251 2.5 kg Plastic bottle 2.5kg
10308310 100 g Plastic bottle 100g
10771614 500 g Plastic bottle 500g
15119253 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11924251 Supplier Thermo Scientific Acros Supplier No. 217520025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7778-18-9 | CaO4S | 136.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
Synonym calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrous
PubChem CID 24497
ChEBI CHEBI:31346
IUPAC Name calcium;sulfate
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1450.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 98.5% min
Linear Formula CaSO4
Fieser 01,107
Merck Index 15,1708
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in glycerol, practically insoluble in most common organic, solvents
Formula Weight 136.14
Percent Purity 99%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Calcium sulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.