missing translation for 'onlineSavingsMsg'
Learn More

Calcium stearate

Product Code. 11328208
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328208 1 kg 1kg
11318208 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328208 Supplier Thermo Scientific Alfa Aesar Supplier No. 039423.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Waterproofing and release agent, stabilizer for PVC, lubricant, conditioning agent | CAS: 1592-23-0 | C36H70CaO4 | 607.03 g/mol

Used as a stabilizer and a lubricant in rigid PVC processing, PVC compounding and moulding, in leather cloth manufacturing. It is used as a lubricant, in iron wire drawing and used as an anti caking agent in detergent Cakes and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1592-23-0
Molecular Formula C36H70CaO4
Molecular Weight (g/mol) 607.03
MDL Number MFCD00036390
InChI Key CJZGTCYPCWQAJB-UHFFFAOYSA-L
Synonym calcium stearate, calcium octadecanoate, flexichem, calcium distearate, octadecanoic acid, calcium salt, aquacal, calstar, flexichem cs, stavinor 30, stearates
PubChem CID 15324
IUPAC Name calcium;octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Ca+2]

Specifications

Melting Point 179°C to 180°C
Quantity 1 kg
Merck Index 14,1703
Solubility Information Soluble in hot pyridine,slightly soluble in oil,but insoluble in alcohol and ether.
Formula Weight 607.04
Physical Form Powder
Chemical Name or Material Calcium stearate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.