missing translation for 'onlineSavingsMsg'
Learn More

Calcium stearate, 5.9 to 7.1% (Ca), powder, Thermo Scientific Chemicals

Product Code. 13585040 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13585040 1 kg 1kg
13595040 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13585040 Supplier Thermo Scientific Acros Supplier No. 449720010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1592-23-0
Molecular Formula C36H70CaO4
Molecular Weight (g/mol) 607.02
MDL Number MFCD00036390
InChI Key CJZGTCYPCWQAJB-UHFFFAOYSA-L
Synonym calcium stearate, calcium octadecanoate, flexichem, calcium distearate, octadecanoic acid, calcium salt, aquacal, calstar, flexichem cs, stavinor 30, stearates
PubChem CID 15324
IUPAC Name calcium;octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Ca+2]

Especificaciones

Melting Point 145°C to 160°C
Physical Form Powder
Quantity 1 kg
Merck Index 15,1705
Solubility Information Solubility in water: insoluble
Formula Weight 607.02
Percent Purity 6.4 to 7.4% (Ca)
Packaging Plastic Bottle
Chemical Name or Material Calcium stearate, 6.4 to 7.4% (Ca)

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.