missing translation for 'onlineSavingsMsg'
Learn More

Calcium standard solution, for AAS, 1 mg/ml Ca in 2 to 5% HNO3

Product Code. 10296390 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10296390

missing translation for 'mfr': Thermo Scientific Acros 195881000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 7440-70-2 | Ca | 40.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium standard solution
Name Note 1 mg/ml Ca in 0.5N HNO3
Packaging Plastic bottle
CAS 7440-70-2, 7697-37-2, 7732-18-5
Physical Form Solution
Molecular Formula Ca
Concentration 997 to 1003 μg/mL of Ca(2+) in 0.5N HNO3 (20°C)
MDL Number MFCD00010897 MFCD00085314
Solubility Soluble in water
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
SMILES [Ca]
IUPAC Name calcium
Molecular Weight (g/mol) 40.08
PubChem CID 5460341
Quantity 100 mL
ChEBI CHEBI:29320
Formula Weight 40.08
Grade AAS
Show More Show Less
compliance-icons
Product Identifier
  • Calcium atomic absorption standard solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.